Learn More
Gibco™ OneFeed™ Supplement
Product Code. 16595601
Click to view available options
Quantity:
2 L
10 L
50 L
Unit Size:
10 L
2 L
50 L
Product Code. 16595601
Brand: Gibco™ 670108
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
A versatile medium supplement.
Gibco™ OneFeed™ Supplement is a chemically-defined, animal-free, protein-free feed shown to enhance protein production when used in a variety of commercially available medium.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Storage conditions: 2°C to 8°C. Store in cool, dry conditions in well-sealed containers.
Cell Line CHO DG44, BHK-21, CHO-K1, CHO
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Powder
Product Type Media Supplement
Format BPC
Quantity 50 L
Source Animal origin-free