missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ONECUT3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568888
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Mouse: 100%; Rat: 93%.
Transcriptional activator. Binds the consensus DNA sequence 5'-DHWATTGAYTWWD-3' on a variety of gene promoters such as those of HNF3B and TTR.
Specifications
| ONECUT3 | |
| Polyclonal | |
| Unconjugated | |
| ONECUT3 | |
| OC3; OC-3; One cut domain family member 3; one cut domain, family member 3; one cut homeobox 3; Onecut3; Transcription factor ONECUT-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 246086 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8K557 | |
| ONECUT3 | |
| synthetic peptide directed towards the c terminal region of mouse ONECUT3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction