missing translation for 'onlineSavingsMsg'
Learn More

Omtriptolide, MedChemExpress

Product Code. 16487038
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487038 10 mg 10mg
16439587 5 mg 5mg
16431609 1 mg 1mg
16423526 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487038 Supplier Medchem Express Supplier No. HY16363
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Omtriptolide (PG490-88) is a derivative prodrug of triptolide purified from the Chinese herb.

Omtriptolide (PG490-88) is a derivative prodrug of triptolide purified from the Chinese herb.

Specifications

Chemical Name or Material Omtriptolide
CAS 195883-06-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H28O9
Quantity 10 mg
Solubility Information H2O : 1.39 mg/mL (3.02 mM; Need ultrasonic)
SMILES C[C@@]([C@@]1([H])C[C@@H]2O3)(CCC4=C1COC4=O)[C@]5([C@H]([C@H]6[C@]7(C(C)C)O6)O5)[C@]23[C@@H]7OC(CCC(O)=O)=O
Molecular Weight (g/mol) 460.47
Formula Weight 460.47
Percent Purity 98.23%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.