missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Omipalisib
GREENER_CHOICE

Product Code. p-7166800
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18715696 10 mg 10mg
18765616 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18715696 Supplier Tocris Bioscience™ Supplier No. 6792/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent PI 3-kinase inhibitor; also potent inhibitor of mTOR and DNA-PK

Omipalisib is a highly potent PI 3-K inhibitor (IC50 = 0.04 nM). Also inhibits mTOR (Ki = 0.18 - 0.3 nM) and DNA-PK (IC50 = 0.28 nM). Induces G1 cell cycle arrest and inhibits proliferation of some breast cancer cells lines in vitro. Reduces pAKT-S473 levels in and inhibits growth of BT474 breast cancer xenografts in mice. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Difluoro-N-[2-methoxy-5-[4-(4-pyridazinyl)-6-quinolinyl]-3-pyridinyl]benzenesulfonamide
Synonym GSK2126458
Quantity 10 mg
Molecular Formula C25H17F2N5O3S
Formula Weight 505.5
Product Type Inhibitor
Purity 98%
Product Title
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.