Learn More
Omeprazole-d3 Sulfone (benzimidazole-4,6,7-d3), CDN
Specifications
Specifications
| Chemical Name or Material | Omeprazole Sulfone D3 (benzimidazole-4,6,7-D3) |
| CAS | 2749628-17-7 |
| Molecular Formula | C17 D3 H16 N3 O4 S |
| Quantity | 0.005 g |
| Synonym | 2-[(4-Methoxy-3,5-dimethylpyridin-2-yl)methylsulfonyl]-5-methoxy-4,6,7-trideuterio-1 H-benzimidazole, 6-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]sulfonyl]-1 H-benzimidazole-4,5,7-d3, Omeprazole-d3 sulfone, Omeprazole sulfone D3 (benzimidazole-4,6,7-D3), Omeprazole-d3 sulfone (benzimidazole-4,6,7-d3) |
| InChI Formula | InChI=1 S/C17H19N3O4S/c1-10-8-18-15(11(2)16(10)24-4)9-25(21,22)17-19-13-6-5-12(23-3)7-14(13)20-17/h5-8 H,9H2,1-4H3,(H,19,20)/i5D,6 D,7 D |
| SMILES | [2 H]c1c([2 H])c2[nH]c(nc2c([2 H])c1OC)S(=O)(=O)Cc3ncc(C)c(OC)c3C |
| IUPAC Name | 4,6,7-trideuterio-5-methoxy-2-[(4-methoxy-3,5-dimethylpyridin-2-yl)methylsulfonyl]-1 H-benzimidazole |
| Molecular Weight (g/mol) | 364.434 |
| Formula Weight | 364.128 g/mol |
| Show More |