missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OMD Monoclonal Antibody (806001)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561219
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
OMD may be implicated in biomineralization processes. This protein has a function in binding of osteoblasts via the alpha(V)beta(3)-integrin.
Specifications
| OMD | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q99983 | |
| OMD | |
| HEK293 human embryonic kidney cell line transfected with human Osteoadherin/OSAD, Gln21-Gln421. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry | |
| 806001 | |
| Unconjugated | |
| OMD | |
| Keratan sulfate proteoglycan osteomodulin; KSPG osteomodulin; Omd; OSAD; Osteoadherin; osteoadherin proteoglycan; osteomodulin; SLRR2C; UNQ190/PRO216; UNQ190/PRO216SLRR2C | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4958 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction