Learn More
Olutasidenib, MedChemExpress
Product Code. 16495596
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16495596
Brand: Medchem Express HY114226
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Olutasidenib (FT-2102) is a highly potent, orally active, brain penetrant and selective inhibitor of mutant Isocitrate dehydrogenase 1 (IDH1), with IC50 values of 21.2 nM and 114 nM for IDH1- R132H and IDH1- R132C, respectively . Olutasidenib (FT-2102) is under the study in the treatment of acute myeloid leukemia (AML) or myelodysplastic syndrome (MDS) .
Olutasidenib (FT-2102) is a highly potent, orally active, brain penetrant and selective inhibitor of mutant Isocitrate dehydrogenase 1 (IDH1), with IC50 values of 21.2 nM and 114 nM for IDH1- R132H and IDH1- R132C, respectively . Olutasidenib (FT-2102) is under the study in the treatment of acute myeloid leukemia (AML) or myelodysplastic syndrome (MDS) .
Specifications
Chemical Name or Material Olutasidenib
Physical Form Solid
CAS 1887014-12-1
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H15ClN4O2
For Use With (Application) Cancer-programmed cell death
Synonym FT-2102
Solubility Information DMSO : 125 mg/mL (352.32 mM; Need ultrasonic)
SMILES N#CC1=CC=C(N[C@H](C2=CC3=C(NC2=O)C=CC(Cl)=C3)C)C(N1C)=O
Molecular Weight (g/mol) 354.79
Formula Weight 354.79
Percent Purity 99.3%
Grade Research