Learn More
Oleuropein, TRC
Product Code. 17959834
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17959834 10 mg 10mg
17948974 25 mg 25mg
17917914 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17959834 Supplier Toronto Research Chemicals Supplier No. TRCO53295110MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32619-42-4
Molecular Formula C12 H17 O7
Molecular Weight (g/mol) 540.51
Synonym (2S,3E,4S)-3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2H-Pyran-4-acetic acid 2-(3,4-dihydroxyphenyl)ethyl ester, 2H-Pyran-4-acetic acid, 3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-, 2-(3,4-dihydroxyphenyl)ethyl ester, [2S-(2α,3E,4β)]-, 2H-Pyran-4-acetic acid, 5-carboxy-3-ethylidene-2-(β-D-glucosyloxy)-3,4-dihydro-, 3,4-dihydroxyphenethyl 5-methyl ester (7CI), Oleuropein (8CI), Oleoeuropein, Oleoeuropeine, Oleuropeine
IUPAC Name methyl (4S,5E,6S)-4-[2-[2-(3,4-dihydroxyphenyl)ethoxy]-2-oxoethyl]-5-ethylidene-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4H-pyran-3-carboxylate
SMILES COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)\[C@@H]1CC(=O)OCCc3ccc(O)c(O)c3
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C25H32O13/c1-3-13-14(9-19(29)35-7-6-12-4-5-16(27)17(28)8-12)15(23(33)34-2)11-36-24(13)38-25-22(32)21(31)20(30)18(10-26)37-25/h3-5,8,11,14,18,20-22,24-28,30-32H,6-7,9-10H2,1-2H3/b13-3+/t14-,18+,20+,21-,22+,24-,25-/m0/s1
Formula Weight 540.1843
Percent Purity >95%
Chemical Name or Material Oleuropein
Grade HPLC