Learn More
Oleuropein (>75%), TRC
Product Code. 30005058
Change view
Click to view available options
Quantity:
25 mg
500 mg
10 g
Unit Size:
10g
25mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005058 10 g 10g
30009140 500 mg 500mg
30007654 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30005058 Supplier Toronto Research Chemicals Supplier No. TRCO53295010G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32619-42-4
Molecular Formula C12 H17 O7
Molecular Weight (g/mol) 540.51
Synonym (2S,3E,4S)-3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-2H-Pyran-4-acetic acid 2-(3,4-dihydroxyphenyl)ethyl ester, 2H-Pyran-4-acetic acid, 3-ethylidene-2-(β-D-glucopyranosyloxy)-3,4-dihydro-5-(methoxycarbonyl)-, 2-(3,4-dihydroxyphenyl)ethyl ester, [2S-(2α,3E,4β)]-, 2H-Pyran-4-acetic acid, 5-carboxy-3-ethylidene-2-(β-D-glucosyloxy)-3,4-dihydro-, 3,4-dihydroxyphenethyl 5-methyl ester (7CI), Oleuropein (8CI), Oleoeuropein, Oleoeuropeine, Oleuropeine
IUPAC Name methyl (4S,5E,6S)-4-[2-[2-(3,4-dihydroxyphenyl)ethoxy]-2-oxoethyl]-5-ethylidene-6-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4H-pyran-3-carboxylate
SMILES COC(=O)C1=CO[C@@H](O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)\C(=C\C)\[C@@H]1CC(=O)OCCc3ccc(O)c(O)c3
Specifications
Quantity 10 g
InChI Formula InChI=1S/C25H32O13/c1-3-13-14(9-19(29)35-7-6-12-4-5-16(27)17(28)8-12)15(23(33)34-2)11-36-24(13)38-25-22(32)21(31)20(30)18(10-26)37-25/h3-5,8,11,14,18,20-22,24-28,30-32H,6-7,9-10H2,1-2H3/b13-3+/t14-,18+,20+,21-,22+,24-,25-/m0/s1
Formula Weight 540.1843
Chemical Name or Material Oleuropein