missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Oleuropein
Description
- Oleuropein is the primary phenolic compound found in olives and olive oil, and it exhibits antioxidant, anti-inflammatory, and anti-cancer properties (Sun et al.)
- Decreases adipocyte differentiation and increases osteoblast differentiation in mesenchymal stem cells (Santiago-Mora et al.)
- Inhibits proliferation and induces apoptosis in MCF-7 human breast cancer cells (Han et al.)
Specifications
Specifications
| CAS | 32619-42-4 |
| Quantity | 100 mg |
| Molecular Formula | C25H32O13 |
| Purity | ≥98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?