missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Olamkicept Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA554954
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Olamkicept acts as an IL-6 trans-signaling inhibitor by selectively binding to the soluble IL-6 receptor (sIL-6R), preventing the formation of the IL-6/sIL-6R complex and subsequent activation of the gp130 receptor, thereby inhibiting downstream inflammatory signaling pathways.
Specifications
| Olamkicept Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| IFNB2IL6R | |
| Recombinant Human IL6 and CD126/IL6R/IL-6RA. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1.3 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction