Learn More
Ohaus™ OIML Class F1 Weights with Certificate
Product Code. 15498934 Shop All Ohaus Products
Click to view available options
:
Weight: 0.194 kg
This item is not returnable. View return policy
Product Code. 15498934
Brand: Ohaus™ 11117096
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
OHAUS™ OIML Class F1 Weights are adjusted to meet or exceed OIML F1 tolerances.
  • Constructed of stainless steel
  • Weights include a weight case
  • Single body design without adjusting cavity
TRUSTED_SUSTAINABILITY
Specifications
Classification OIML F1 Weights
Weight (Metric) 0.194 kg
Includes Certificate