missing translation for 'onlineSavingsMsg'
Learn More

OGT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16808753 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16808753 150 μL 150µL
16818753 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16808753 Supplier Proteintech Supplier No. 115762AP150UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

O-linked N-acetylglucosamine (O-GlcNAc), is a member of the O-GlcNAc transferase family. O-GlcNAc is a posttranslational modification characterized by the attachment of N-acetylglucosamine to specific serine/threonine hydroxyl groups. It plays an important role in nutrient sensing, gene expression, and protein degradation.

Specifications

Antigen OGT
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene OGT
Gene Accession No. O15294, P56558
Gene Alias HRNT1, O GLCNAC, OGT
Gene Symbols OGT
Host Species Rabbit
Immunogen OGT Fusion Protein Ag2160
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108155, 26295, 8473
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.