Learn More
Tocris Bioscience™ OGT 2115
GREENER_CHOICE
Product Code. 18045912
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18045912 1 mg 1mg
18024112 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18045912 Supplier Tocris Bioscience™ Supplier No. 2710/1
Stock Up and Save This Summer
Use promo code 27950
View all promotions
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Antiangiogenic. Heparanase inhibitor
OGT 2115 is a heparanase inhibitor (IC50 = 0.4 μM) that displays no major inhibition of human cytochrome P450 isoenzymes (IC50 > 30 μM). Exhibits antiangiogenic properties in vitro (IC50 = 1 μM).
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-[4-[[3-(4-Bromophenyl)-1-oxo-2-propenyl]amino]-3-fluorophenyl]-5-benzoxazoleacetic acid
CAS 853929-59-6
Assay Percent Range >98%
Quantity 1 mg
Solubility Soluble to 10mM in DMSO
Molecular Formula C24H16BrFN2O4
Formula Weight Observed MW: 495.3
Purity 97%