Learn More
Novus Biologicals™ OGR1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30855-100ug
 View more versions of this product
Product Code. 30067206
  • 861.00 € / 100µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
OGR1 Polyclonal antibody specifically detects OGR1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OGR1
Polyclonal
Western Blot : 5-10 μg/mL, Immunohistochemistry, Functional
G protein-coupled receptor 68, GPR12A, G-protein coupled receptor 68, OGR-1, ovarian cancer G protein-coupled receptor, 1, ovarian cancer G-protein coupled receptor 1, Sphingosylphosphorylcholine receptor
Rabbits were immunized with recombinant protein derived from human OGR1 (GPR68).
100 μg
GPCR
8111
Store at -20°C. Avoid freeze/thaw cycles.
IgG
Immunohistochemistry, Western Blot, Functional Assay
Unconjugated
Phosphate buffered saline
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified