missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ODF4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA543975

Product Code. 15516302

  • 530.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: MDAEYSGNEF PRSEGERDQH QRPGKERKSG EAGWGTGELG QDGRLLSSTL Sequence homology: Human: 100%.

ODF4 is the component of the outer dense fibers (ODF) of spermatozoa which could be involved in sperm tail structure, sperm movement and general organization of cellular cytoskeleton.This gene encodes a protein that is localized in the outer dense fibers of the tails of mature sperm. This protein is thought to have some important role in the sperm tail. Sequence Note: The RefSeq record was created from transcript and genomic sequence data to make the sequence consistent with the reference genome assembly. The genomic coordinates used for the transcript record were based on transcript alignments.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ODF4
Polyclonal
Unconjugated
ODF4
cancer/testis antigen 134; cancer/testis antigen 136; CT134; CT136; hOPPO1; MGC138215; MGC138219; ODF4; OPPO 1; OPPO1; outer dense fiber 4; outer dense fiber of sperm tails 4; outer dense fiber of sperm tails protein 4; Outer dense fiber protein 4; testis-specific protein oppo 1
Rabbit
Affinity chromatography
RUO
146852
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q2M2E3
ODF4
Synthetic peptide directed towards the N-terminal of human ODF4 (aa 1-50).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.