Learn More
Octahydrocyclopenta[b]morpholine, TRC
Product Code. 30411927
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30411927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30411927 Supplier Toronto Research Chemicals Supplier No. TRCO30766650MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Octahydrocyclopenta[b]morpholine
CAS 1018639-83-2
Molecular Formula C7 H13 N O
Quantity 50 mg
Synonym Octahydrocyclopent[b]-1,4-oxazine, 2,3,4,4a,5,6,7,7a-Octahydrocyclopenta[b][1,4]oxazine, Octahydrocyclopenta[b][1,4]oxazine
InChI Formula InChI=1S/C7H13NO/c1-2-6-7(3-1)9-5-4-8-6/h6-8H,1-5H2
SMILES C1CC2NCCOC2C1
IUPAC Name 2,3,4,4a,5,6,7,7a-octahydrocyclopenta[b][1,4]oxazine
Molecular Weight (g/mol) 127.184
Formula Weight 127.1