Learn More
BRAND™ Octagonal Shape PTFE Magnetic Stirring Bars
PTFE construction. Angular surface creates high turbulence even at low speeds.
45.41 € - 142.00 €
Specifications
Material | PTFE |
---|---|
Shape | Octagon |
For Use With (Equipment) | For vessels with slightly uneven bottoms |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11804100
|
BRAND™
137625 |
12 mm | 75 mm |
142.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
12030932
|
BRAND™
137607 |
8 mm | 13 mm |
45.41 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
12040932
|
BRAND™
137608 |
8 mm | 15 mm |
48.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11844090
|
BRAND™
137610 |
9.5 mm | 25 mm |
48.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
12055282
|
BRAND™
137612 |
11 mm | 25 mm |
52.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
12065282
|
BRAND™
137617 |
11 mm | 38 mm |
76.55 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
12075282
|
BRAND™
137620 |
11 mm | 51 mm |
76.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
11894090
|
BRAND™
137622 |
11 mm | 64 mm |
95.50 €
Pack of 10 |
In Stock
Log in to see stock. |
||||
10191631
|
BRAND™
137618 |
9.5 mm | 51 mm |
70.00 €
Pack of 10 |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
Description
- Octagonal shape
- For vessels with slightly uneven bottoms.
- Pivot ring reduces the contact surface which enables the stirrer to adopt the optimum position.
Specifications
PTFE | |
Octagon | |
For vessels with slightly uneven bottoms |