Learn More
Vitlab™ Octagonal Magnetic Stir Bar
Vitlab™ Octagonal Magnetic Stir Bar with rings and permanent magnet AlNiCo V core.
Brand:  Vitlab™ 308097
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10595021
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • The eight-sided shape gives rise to significant turbulence, and thus achieves effective mixing, even at low spin rates
  • The middle ring also promotes stable centering with convex or uneven bottoms
Specifications
Specifications
8 mm
28 mm
Octagon