Learn More
Vitlab™ Octagonal Magnetic Stir Bar
Vitlab™ Octagonal Magnetic Stir Bar with rings and permanent magnet AlNiCo V core.
Brand: Vitlab™ 308097
Additional Details : Weight : 0.01000kg
Description
- The eight-sided shape gives rise to significant turbulence, and thus achieves effective mixing, even at low spin rates
- The middle ring also promotes stable centering with convex or uneven bottoms
Specifications
8 mm | |
28 mm | |
Octagon |