missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OCT4 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518403
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
POU5F1, also commonly known as Oct-4, is a maternally expressed octamer-binding protein that was the first transcription factor described for the early stages of development. The role of POU5F1 in embryonic development suggested that it might be useful in the creation of stem cells that might be useful in cell replacement therapies in the treatment of several degenerative diseases. Artificial stem cells, termed induced pluripotent stem (iPS) cells, can be created by expressing POU5F1 and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing POU5F1, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine.
Specifications
| OCT4 | |
| Polyclonal | |
| Unconjugated | |
| POU5F1 | |
| 2-Oct; 3-Oct; 4-Oct; cb197; chunp6868; DADB-104B20.2; etID49452.21; gp9; gp-9; HGNC:9221; homeobox protein; lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; MGC22487; NF-A3; Oct2; Oct-2; Oct2a; Oct2b; OCT3; Oct-3; Oct3/4; Oct-3/4; OCT4; Oct-4; Octamer-binding protein 2; octamer-binding protein 3; Octamer-binding protein 4; octamer-binding transcription factor 2; Octamer-binding transcription factor 3; OTF2; OTF-2; OTF3; Otf-3; Otf3g; Otf3-rs7; Otf4; Otf-4; OTTHUMP00000221150; OTTHUMP00000221151; POU class 2 homeobox 2; POU class 5 homeobox 1; POU class 5 homeobox 1 transcript variant OCT4B1; POU class 5 homeobox 1 transcript variant OCT4B2; POU class 5 homeobox 1 transcript variant OCT4B3; POU class 5 homeobox 1 transcript variant OCT4B4; POU class 5 homeobox 1 transcript variant OCT4B5; POU class 5 homeobox 1 transcript variant OCT4B6; POU domain class 5 transcription factor 1; POU domain gene 2; POU domain protein 2; POU domain transcription factor OCT4; POU domain, class 2, transcription factor 2; POU domain, class 5, transcription factor 1; POU domain, class 5, transcription factor 3; pou2; pou-2; POU2F2; Pou5f1; pou5f1 protein; pou5f3; POU-type homeodomain-containing DNA-binding protein; spg; spg/pou2; spiel-ohne-grenzen; spiel-ohne-grenzen/pou2; wu:fd18d06 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 18999, 5460 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P20263, Q01860 | |
| POU5F1 | |
| Synthetic peptide sequence (VTTLGSPMHSN) corresponding to the C-terminus amino acids of POU5F1 (aa 350-360). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction