Learn More
Motic™ Objective, Plan Phase PL Ph 40X/0.65/S
Click to view available options
Magnification Power:
40X
Unit Size:
Each
Description
- Plan style for field curvature correction.
- Integrated phase plate for contrast modulation.
Specifications
Specifications
| Type | Objective |
| Description | NA: 0.65/S |
| For Use With (Equipment) | B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes |
| Magnification Power | 40X |