Learn More
Obatoclax Mesylate, MedChemExpress
Product Code. 16460647
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16460647
Brand: Medchem Express HY10969
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Obatoclax Mesylate (GX15-070 Mesylate), a BH3 mimetic, is a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. Obatoclax Mesylate induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. Obatoclax Mesylate has anti-cancer and broad-spectrum antiparasitic activity.
Obatoclax Mesylate (GX15-070 Mesylate), a BH3 mimetic, is a pan-BCL-2 family proteins inhibitor with a Ki of 220 nM for BCL-2. Obatoclax Mesylate induces autophagy-dependent cell death and targets cyclin D1 for proteasomal degradation. Obatoclax Mesylate has anti-cancer and broad-spectrum antiparasitic activity.
Specifications
Chemical Name or Material Obatoclax Mesylate
Physical Form Solid
CAS 803712-79-0
Color Black
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H23N3O4S
For Use With (Application) Cancer-Kinase/protease
Synonym GX15-070 Mesylate
Solubility Information DMSO : 12.5 mg/mL (30.23 mM; Need ultrasonic)
SMILES COC1=CC(C2=CC3=C(C=CC=C3)N2)=N/C1=C\C4=C(C=C(N4)C)C.CS(=O)(O)=O
Molecular Weight (g/mol) 413.49
Formula Weight 413.49
Percent Purity 98.0%
Grade Research