Learn More
O3-Desethyl Apremilast, TRC
Specifications
Specifications
| Chemical Name or Material | O3-Desethyl Apremilast |
| CAS | 1384967-20-7 |
| Molecular Formula | C20H20N2O7S |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C20H20N2O7S/c1-11(23)21-14-6-4-5-13-18(14)20(26)22(19(13)25)15(10-30(3,27)28)12-7-8-17(29-2)16(24)9-12/h4-9,15,24H,10H2,1-3H3,(H,21,23)/t15-/m0/s1 |
| SMILES | OC1=CC([C@@H](N2C(C(C=CC=C3NC(C)=O)=C3C2=O)=O)CS(=O)(C)=O)=CC=C1OC |
| Molecular Weight (g/mol) | 432.45 |
| Formula Weight | 432.0991 |