Learn More
Alfa Aesar™ O-(N-Succinimidyl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate, 98+%
Product Code. 15465265
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15465265 1 g 1g
15475265 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15465265 Supplier Alfa Aesar™ Supplier No. H54371.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 207683-26-9
Molecular Formula C13H20F6N3O3P
Molecular Weight (g/mol) 411.29
MDL Number MFCD00192457
InChI Key TVWKVGNCIJUXMY-UHFFFAOYSA-N
Synonym hspyu, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate, n,n,n',n'-bis tetramethylene-o-n-succinimidyl uronium hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 2,5-dioxopyrrolidin-1-yl oxy bis pyrrolidin-1-yl methylium hexafluorophosphate, dipyrrolidino n-succinimidyloxy carbenium hexafluorophosphate chn, bis pyrrolidin-1-yl methylidene 2,5-dioxopyrrolidin-1-yl oxidanium hexafluorophosphate, 1-1-pyrrolidin-1-iumylidene 1-pyrrolidinyl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-pyrrolidin-1-ium-1-ylidene pyrrolidin-1-yl methoxy pyrrolidine-2,5-dione hexafluorophosphate, 1-2,5-dioxopyrrolidin-1-yl oxy pyrrolidin-1-yl methylidene-1??-pyrrolidin-1-ylium hexafluorophosphate
PubChem CID 16218611
IUPAC Name 1-{[(2,5-dioxopyrrolidin-1-yl)oxy](pyrrolidin-1-yl)methylidene}-1λ⁵-pyrrolidin-1-ylium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.O=C1CCC(=O)N1OC(N1CCCC1)=[N+]1CCCC1
Specifications
Melting Point 152°C to 154°C
Quantity 1 g
Formula Weight 411.28
Percent Purity ≥98%
Chemical Name or Material O-(N-Succinimidyl)-N,N,N',N'-bis(tetramethylene)uronium hexafluorophosphate