Learn More
O,N-Aminomethanylylidene-beta-D-arabinofuranose, TRC
Specifications
Specifications
| Chemical Name or Material | O,N-Aminomethanylylidene-beta-D-arabinofuranose |
| CAS | 27963-98-0 |
| Molecular Formula | C6 H10 N2 O4 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C6H10N2O4/c7-6-8-5-4(12-6)3(10)2(1-9)11-5/h2-5,9-10H,1H2,(H2,7,8)/t2-,3-,4+,5-/m1/s1 |
| SMILES | NC1=N[C@@H]2O[C@H](CO)[C@@H](O)[C@@H]2O1 |
| IUPAC Name | (3aR,5R,6R,6aS)-2-amino-5-(hydroxymethyl)-3a,5,6,6a-tetrahydrofuro[2,3-d][1,3]oxazol-6-ol |
| Molecular Weight (g/mol) | 174.15 |
| Formula Weight | 174.0641 |