Learn More
o-IDTBr, Thermo Scientific Chemicals
Product Code. 17126574
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17126574 500 mg 500mg
17106574 100 mg 100mg
17116574 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17126574 Supplier Thermo Scientific Alfa Aesar Supplier No. H66666.ME
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2077945-91-4
Molecular Formula C72H88N6O2S8
Molecular Weight (g/mol) 1326.02
InChI Key HTXQPYFGYQYREE-UHFFFAOYSA-N
Synonym 5,​5 '-​[(4,​9-​dihydro-​4,​4,​9,​9-​tetraoctyl-​s-​indaceno[1,​2-​b:5,​6-​b']​dithiophene-​2,​7-​diyl)​bis(2,​1,​3-​benzothiadiazole-​7,​4-​diylmethylidyne)​]​bis[3-​ethyl-​2-​thioxo-4-​Thiazolidinone
IUPAC Name 3-ethyl-5-{[7-(15-{7-[(3-ethyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene)methyl]-2,1,3-benzothiadiazol-4-yl}-9,9,18,18-tetraoctyl-5,14-dithiapentacyclo[10.6.0.0³,¹⁰.0⁴,⁸.0¹³,¹⁷]octadeca-1(12),2,4(8),6,10,13(17),15-heptaen-6-yl)-2,1,3-benzothiadiazol-4-yl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
SMILES CCCCCCCCC1(CCCCCCCC)C2=C(SC(=C2)C2=CC=C(C=C3SC(=S)N(CC)C3=O)C3=NSN=C23)C2=CC3=C(C=C12)C1=C(C=C(S1)C1=CC=C(C=C2SC(=S)N(CC)C2=O)C2=NSN=C12)C3(CCCCCCCC)CCCCCCCC
Specifications
Quantity 500 mg
Formula Weight 1326.03
Physical Form Solid
Chemical Name or Material o-IDTBr