Learn More
O-Acetyl Phenylephrine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | O-Acetyl Phenylephrine Hydrochloride |
| CAS | 2732261-25-3 |
| Molecular Formula | C11H15NO3 •HCl |
| Quantity | 100 mg |
| Synonym | 1-Acetate-3-hydroxy-α-[(methylamino)methyl]-(R)-benzenemethanol Hydrochloride |
| InChI Formula | InChI=1S/C11H15NO3.ClH/c1-8(13)15-11(7-12-2)9-4-3-5-10(14)6-9;/h3-6,11-12,14H,7H2,1-2H3;1H/t11-;/m0./s1 |
| SMILES | OC1=CC=CC([C@@H](OC(C)=O)CNC)=C1.Cl |
| IUPAC Name | (R)-1-(3-hydroxyphenyl)-2-(methylamino)ethyl acetate hydrochloride |
| Molecular Weight (g/mol) | 245.7 |
| Formula Weight | 245.09 |