missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NXF1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06455H
This item is not returnable.
View return policy
Description
NXF1 Polyclonal antibody specifically detects NXF1 in Human samples. It is validated for Western Blot
Specifications
| NXF1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10482 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| DKFZp667O0311, Mex67, mRNA export factor TAP, nuclear RNA export factor 1, TAPMEX67, tip associating protein, Tip-associated protein, Tip-associating protein | |
| Produced in rabbits immunized with E. coli-derived Human NXF1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction