Learn More
NVP-HSP990, MedChemExpress
Product Code. 16462467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462467 50 mg 50mg
16426358 10 mg 10mg
16418947 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16462467 Supplier Medchem Express Supplier No. HY15190
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress NVP-HSP990 is a potent and selective Hsp90 inhibitor, with IC50 values of 0.6, 0.8, and 8.5 nM for Hsp90α, Hsp90β, and Grp94, respectively.
NVP-HSP990 is a potent and selective Hsp90 inhibitor, with IC50 values of 0.6, 0.8, and 8.5 nM for Hsp90α, Hsp90β, and Grp94, respectively.
Specifications
Chemical Name or Material NVP-HSP990
CAS 934343-74-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H18FN5O2
Quantity 50 mg
Synonym HSP-990
Solubility Information DMSO : ≥ 33 mg/mL (86.98 mM)
SMILES CC1=C2C(C[C@H](C3=C(C4=NC(OC)=CC=C4)C=C(F)C=C3)NC2=O)=NC(N)=N1
Molecular Weight (g/mol) 379.39
Formula Weight 379.39
Percent Purity 97.2%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease