Learn More
NUSAP1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Product Code. 18074805 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18074805 20 μL 20µL
18064515 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18074805 Supplier Proteintech Supplier No. CL4881202420UL
Please to purchase this item. Need a web account? Register with us today!
Rabbit Polyclonal Antibody
The NUSAP1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NUSAP1. This antibody recognizes human antigen. The NUSAP1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Antigen NUSAP1
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols ANKT, BM037, FLJ13421, LNP, NuSAP, NUSAP1, PRO0310, PRO0310p1, Q0310, SAPL
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 51203
Target Species Human
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG