Learn More
NUPR1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15056-1-AP-20UL
 View more versions of this product
Product Code. 16804044
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
The human p8 gene encodes a nuclear protein with strong mitogenic activity, which is expressed in the pancreas in response to acute pancreatitis. Response to TNF alpha and staurosporine-induced apoptosis is not modified by p8 expression. However, cells expressing p8 grow significantly and more rapidly.
Specifications
Specifications
NUPR1
Polyclonal
Unconjugated
NUPR1
Candidate of metastasis 1, COM1, nuclear protein 1, NUPR1, P8, Protein p8
Rabbit
Antigen Affinity Chromatography
RUO
26471
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60356
NUPR1
NUPR1 Fusion Protein Ag7039