Learn More
Numidargistat dihydrochloride, MedChemExpress
Product Code. 16483668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16483668 10 mg 10mg
16456407 5 mg 5mg
16412826 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16483668 Supplier Medchem Express Supplier No. HY101979A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Numidargistat (CB-1158) dihydrochloride is a potent and orally active inhibitor of arginase, with IC50s of 86 nM and 296 nM for recombinant human arginase 1 and recombinant human arginase 2, respectively. Immuno-oncology agent.
Numidargistat (CB-1158) dihydrochloride is a potent and orally active inhibitor of arginase, with IC50s of 86 nM and 296 nM for recombinant human arginase 1 and recombinant human arginase 2, respectively. Immuno-oncology agent.
Specifications
Chemical Name or Material Numidargistat dihydrochloride
Color White
Purity Grade Notes Research
Molecular Formula C11H24BCl2N3O5
Quantity 10 mg
Synonym CB-1158 dihydrochlorideINCB01158 dihydrochloride
Solubility Information DMSO : 55 mg/mL (152.76 mM; Need ultrasonic) ∣H2O : 37.78 mg/mL (104.93 mM; Need ultrasonic)
SMILES O=C([C@@H](N)C)N1C[C@H](CCCB(O)O)[C@](N)(C(O)=O)C1.Cl.Cl
Molecular Weight (g/mol) 360.04
Formula Weight 360.04
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death