missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NuMA Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598021
This item is not returnable.
View return policy
Description
NuMA Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
NuMA (nuclear mitotic apparatus) is a long coil-coiled protein that plays a role in the interphase and mitosis phases of a cell cycle. In interphase, it has been hypothesized to play a structural role in the nucleoskeleton that may be important in nuclear organization and functions. During mitosis, it is associated with spindle microtubule organization and chromosome positioning.
Specifications
| NuMA | |
| Polyclonal | |
| Unconjugated | |
| NUMA1 | |
| 6720401E04Rik; AA764025; AL022610; AU014979; centrophilin stabilizes mitotic spindle in mitotic cells; NMP22; NMP-22; Nuclear matrix protein-22; Nuclear mitotic apparatus protein; Nuclear mitotic apparatus protein 1; NUMA; NuMA protein; Numa1; SP-H antigen; structural nuclear protein | |
| Rabbit | |
| Protein G | |
| RUO | |
| 4926 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q14980 | |
| NUMA1 | |
| Recombinant Human Nuclear mitotic apparatus protein 1 protein (627-870AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction