Learn More
NUDT6 Monoclonal Antibody (OTI9F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502330S
Description
NUDT6 Monoclonal antibody specifically detects NUDT6 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| NUDT6 | |
| Monoclonal | |
| 0.86 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P25686 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3300 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI9F5 | |
| Unconjugated | |
| DNAJB2 | |
| antisense basic fibroblast growth factor; Antisense basic fibroblast growth factor B; anti-sense basic fibroblast growth factor B; ASFGF2; Asfgf2b; FGF2AS; FGF-AS; Gfg; GFG1; GFG-1; Nucleoside diphosphate-linked moiety X motif 6; nudix (nucleoside diphosphate linked moiety X)-type motif 6; nudix hydrolase 6; nudix motif 6; Nudt6; Protein GFG | |
| Full length human recombinant protein of human DNAJB2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.