missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUDT10 Monoclonal Antibody (OTI2D9), TrueMAB™, OriGene
Description
NUDT10 Monoclonal antibody specifically detects NUDT10 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | NUDT10 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI2D9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | NUDT10 |
| Gene Accession No. | Q8NFP7 |
| Gene Alias | AI853080; APS2; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-alpha; Diadenosine 5',5'''-P1,P6-hexaphosphate hydrolase 3-beta; Diadenosine hexaphosphate hydrolase (AMP-forming); diphosphoinositol polyphosphate phosphohydrolase 3-alpha; Diphosphoinositol polyphosphate phosphohydrolase 3-beta; DIPP3 alpha; Dipp3a; DIPP-3-alpha; DIPP3-alpha; Dipp3b; DIPP-3-beta; hAps2; hDIPP3alpha; MNCb-1696; Nucleoside diphosphate-linked moiety X motif 10; Nucleoside diphosphate-linked moiety X motif 11; nudix (nucleoside diphosphate linked moiety X)-type motif 10; nudix hydrolase 10; nudix motif 10; Nudix motif 11; Nudt10; Nudt11; RGD1564763 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?