missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ Tissue XS, Micro kit for DNA from cells and tissue
Click to view available options
No. of Reactions:
10
250
Unit Size:
10 reactions
250 reactions
Description
Two different lysis buffers plus optional addition of the chemical additive Enhancer SX allow fine tuning for maximum yield and purity. NucleoSpin™ has been designed for Gram ± bacteria, archaea, fungi, and algae in soil, sludge and sediment samples. NucleoSpin™ bead tubes have ceramic beads included, to be used in a bead mill or vortexer. Efficient mechanical pulping even for Gram bacteria and spores. The Inhibitor removal columns and efficient wash buffers will eliminate PCR inhibitors. DNA can mostly be used undiluted as a PCR template. Highest sensitivity even for rare targets.
- Alternative lysis systems - highest yield from diverse samples
- Ceramic beads included - most efficient cell disruption
- Highest purity - complete removal or PCR* inhibitors For purification of genomic DNA from environmental samples.
Specifications
Specifications
| Buffer | Yes |
| Final Product Type | NucleoSpin Tissue XS Column |
| No. of Reactions | 10 |
| Probe Length | 200 to 50bp to kbp |
| Product Type | NucleoSpin Tissue XS Column |
| Sample Volume | 0.025-10 mg human /animal tissue, 1-30 μL whole blood, 10-104 cultured cells, spots of 5-30 mm2 Guthrie cards |
| Yield | 0.1 to 0.5 ng (102 HeLa cells), 1 to 5 ng (103 HeLa cells), 10 to 50 ng (104 HeLa cells), 20 to 100 ng (0.025 mg mouse liver), 200 to 1000 ng (0.25 mg mouse liver), 600 to 3000 ng (2.5 mg mouse liver) |
Safety and Handling
Hazard Category
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction