missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Tissue Columns, Mini columns for DNA from cells and tissue

Product Code. 15860968
Click to view available options
Quantity:
250 columns
50 columns
Unit Size:
Pack of 250
Pack of 50
This item is not returnable. View return policy

Product Code. 15860968

Brand: Macherey-Nagel™ 740952.50S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mini spin kit for purification of DNA from a broad range of samples.

NucleoSpin™ Tissue Columns

  • Separate Columns with high DNA recovery and purity
  • Reliable DNA purification for reproducible results

Specifications

Cell Type Animal & Human tissue, cells
Capacity 60 μg
Column Type Mini spin column
Content And Storage 18°C to 25°C
Description Columns for purification of DNA from a broad range of samples
Detection Method Silica-membrane Technology
Format Mini Spin Column
Isolation Technology Silica Membrane Technology
Sample Type Animal and Human Tissue, Cells
Elution Volume 60 to 100 μL
Final Product Type DNA
For Use With (Equipment) Centrifugation, Vacuum
For Use With (Application) Genotyping, Multiplex PCR, Next Generation Sequencing
Includes NucleoSpin Tissue Columns
Key Functions DNA purification
No. of Reactions 50
Nucleic Acid Type DNA
Product Type DNA purification columns
Quantity 50 columns
Purity A260/A280 1.7 - 1.9
Sample Volume <25 mg human/animal tissue, 102-107 cultured cells
Shipping Condition Room Temperature
Size 200 bp to 50 kbp
Source Animal & Human tissue, cells
Target DNA
Technique Silica-membrane technology
Test Time 20 min/prep (excl. lysis)
Yield 20 to 35 μg
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • NucleoSpin Tissue
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : guanidine hydrochloride / 2-propanol

For research use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.