missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoSpin™ Blood L, Midi kit for DNA from blood using vacuum processing

Product Code. p-200104878
Change view
Click to view available options
Quantity:
100 preps
24 preps
Unit Size:
100 reactions
24 preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15340195 24 preps 24 preps
15845224 100 preps 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15340195 Supplier Macherey-Nagel™ Supplier No. 740954.24

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Complete kit with silica membrane midi spin columns for rapid medium-scale purification of genomic DNA from blood and biological samples.

NucleoSpin™ Blood L

  • Ready-to-use DNA in less than 60 min
  • Complete removal of PCR inhibitors
  • Consistently high DNA yields
  • Suitable for whole blood (human or animal blood, fresh or frozen)
    • DNA from whole blood treated with citrate, EDTA, heparin, CPDA
    • DNA from serum, plasma, buffy coat, platelets, body fluids (e.g., amniotic fluid)
    • DNA from cultured cells
  • Typical downstream applications: PCR, Southern blotting, enzymatic reactions

Specifications

Color White band
Concentration 200 to 500 ng/μL
Content And Storage Columns: 4 °C; Other components: Room temperature (18-25 °C)
Format Midi Spin Column
Isolation Technology Silica Membrane Technology
Sample Type Cell Cultures, Whole Blood
Efficiency Binding capacity: 250 μg
Elution Volume Elution volume: 120 - 200 μL
Final Product Type Genomic DNA
For Use With (Equipment) Centrifuge
For Use With (Application) Medium scale purification of DNA from blood
Includes NucleoSpin Blood L Columns with Collection Tubes, Collection Tubes (15 mL), buffers, Proteinase K
No. of Reactions 24 Preps
Quantity 24 preps
Purity A260/A280: 1.6 - 1.9
Sample Volume 0.2 - 2 mL blood, 2 x 107 cells
Test Time 60 min/prep
Yield 40 - 60 μg (2 mL whole blood)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Proteinase k, propan-2-ol, guanidine hydrochloride, ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.