missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 8 Tissue, 8-well kit for DNA from cells and tissue
Click to view available options
:
No. of Reactions: 12 x 8
Description
NucleoSpin™ 8 Tissue is designed for the rapid simultaneous preparation of highly pure genomic DNA from animal and human tissue, e.g., mouse and rat tails. From a 1.2cm mouse tail tip section (age of mice 4 to 6 weeks) up to 30μg to 40μg of pure genomic DNA can be prepared (typical yields 15μg to 25μg). The obtained DNA can be used directly for PCR*, Southern blotting or any kind of enzymatic reaction. Parallel lysis of large sample numbers can be performed in standard tubes or in 96 well blocks alternatively. NucleoSpin™ Tissue binding strips can be used under vacuum or centrifugation in a centrifuge with deep buckets allowing processing of NucleoSpin™ 8 well strips stacked on 96 well blocks. The method is based on a specially treated silica membrane with a high binding capacity for nucleic acids. The obtained DNA can be used directly for PCR*, solution blotting, or any kind of enzymatic reaction. Kit components: NucleoSpin™ Tissue binding strips, buffers, proteinase K, MN wash plates, MN square-well blocks, racks with tube strips, self-adhering foil. For further details on this product please contact Customer Services.
- Silica membrane technology
- Format: 8 well strip
- Preparation time: 1hr (after lysis)
- Sample material: <20mg tissue or 106 cultured cells, bacteria
- Yield: 15μg to 25μg
- Elution volume: 100μL to 200μL
- DNA binding capacity: 40μg
- Fragment size: 300bp to 30kb
- No use of organic solvents For manual or automated isolation of DNA from human and animal tissue, cultured cells or bacteria.
Specifications
Specifications
| No. of Reactions | 12 x 8 Preps |
| Product Type | 8 Tissue |
| Sample Volume | <20 mg human/animal tissue, <106 cells |
Safety and Handling
Signal Word
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H336-May cause drowsiness or dizziness.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : guanidine hydrochloride,2-propanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction