missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Nucleoprotein (118-126)

Product Code. 16469638
Encompass_Preferred
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16469638

Brand: MedChemExpress HYP1584

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Nucleoprotein (118-126) is a 9-aa peptide, a fragment of Nucleoprotein.

Specifications

CAS 124454-83-7
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 1008.15 g/mol
Product Type Peptides
Quantity 10 mg
Shipping Condition Room temperature
Synonym NP(118-126)
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.