missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoBond™ Xtra BAC

Product Code. 12723401
Change view
Click to view available options
:
25 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12723401 25 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12723401 Supplier Macherey-Nagel™ Supplier No. 740436.25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The second generation of anion exchanger designed for ultra fast purification of very large construct plasmid DNA (P1, BACs, PACs) ranging from 3 kbp up to 300 kbp.

NucleoBond™ Xtra BAC — For transfection-grade, large construct plasmid DNA

  • Optimized column design – one prep in approximately 75 min
  • Column filter included – parallel lysate clearing and loading onto the column
  • Optimal silica material – yields up to 150 μg
  • Transfection-grade plasmid DNA using proven anion-exchange technology
  • LyseControl for visualization of a completed lysis / neutralization
  • Typical downstream applications: cloning, PCR, restriction analysis, sequencing, and transfection

Specifications

Content And Storage Room temperature (18°C to 25°C)
Format Gravity-flow Column
Isolation Technology Anion Exchange Chromatography Technology
Sample Type E. coli Culture
Efficiency Binding capacity: 150 μg
Final Product Type Transfection-grade, large construct plasmid DNA
For Use With (Equipment) Smart Rack, Combi Rack or equivalent holder
For Use With (Application) Large construct DNA purification
Includes NucleoBond Xtra BAC Columns with inserted NucleoBond Xtra Column Filters, buffers, RNase A
No. of Reactions 25 Reactions
Purity A260/A280: 1.80 - 1.95
Sample Volume 250 to 750 mL
Size < 300 kbp
Test Time 75 min./2 to 4 Preps
Yield 10 to 150 μg
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: ethanol (diluted < 20 %) / sodium hydroxide solution (diluted < 2 %), dodecyl sulfate, sodium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.