missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nuclear Pore-O-Linked Glycoprotein Antibody (RL1), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB600-1068
This item is not returnable.
View return policy
Description
Nuclear Pore-O-Linked Glycoprotein Monoclonal specifically detects Nuclear Pore-O-Linked Glycoprotein in Human, Mouse, Rat, Mammal, Xenopus, Yeast samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Nuclear Pore-O-Linked Glycoprotein | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| PEG purified | |
| RUO | |
| Detects nuclear pore-O-linked glycoprotein | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunofluorescence | |
| RL1 | |
| Western Blot 1:1000, Immunohistochemistry 1:200, Immunoprecipitation 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Immunofluorescence 1:100 | |
| Pore complex-lamina fraction purified from rat liver nuclear envelopes. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Mammalia, Xenopus species, Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction