Learn More
NUBPL Monoclonal Antibody (OTI7D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503746S
Description
NUBPL Monoclonal antibody specifically detects NUBPL in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| NUBPL | |
| Monoclonal | |
| 0.75 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q8TB37 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 80224 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Western Blot | |
| OTI7D6 | |
| Unconjugated | |
| Nubpl | |
| 2410170E07Rik; C14orf127; huInd1; IND1; IND1 homolog; Iron-sulfur protein NUBPL; iron-sulfur protein required for NADH dehydrogenase; NUBPL; nucleotide binding protein like; nucleotide binding protein-like; nucleotide-binding protein-like; RGD1307232 | |
| Human recombinant protein fragment corresponding to amino acids 1-250 of human NUBPL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.