Learn More
MedChemExpress NSP-SA-NHS
Click to view available options
:
5 mg
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 681.73 g/mol |
| Solubility | DMSO : 4 mg/mL (5.87 mM; ultrasonic and warming and heat to 60°C) |
| Content And Storage | -20°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 199293-83-9 |