missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NSE2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA565676

Product Code. 15948414

  • 572.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: SQTNFTCPITK EEMKKPVKNK VCGHTYEEDA IVRMIESRQK RKKKAYCPQI GCSHTDIRKS DLIQDEALRR AIENHNKK Highest antigen sequence identity to the following orthologs - mouse 84%, rat 84%.

This gene encodes the large subunit of DNA damage-binding protein which is a heterodimer composed of a large and a small subunit. This protein functions in nucleotide-excision repair. Its defective activity causes the repair defect in the patients with xeroderma pigmentosum complementation group E (XPE). However, it remains for mutation analysis to demonstrate whether the defect in XPE patients is in this gene or the gene encoding the small subunit. In addition, Best vitelliform mascular dystrophy is mapped to the same region as this gene on 11q, but no sequence alternations of this gene are demonstrated in Best disease patients.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NSE2
Polyclonal
Unconjugated
Nsmce2
1110014D18Rik; AI661537; C8orf36; E3 SUMO-protein ligase NSE2; E3 SUMO-protein transferase NSE2; FLJ32440; hMMS21; methyl methanesulfonate sensitivity gene 21; MMS21; MMS21 homolog; non-SMC element 2 homolog; non-SMC element 2 homolog (MMS21, S. cerevisiae); non-SMC element 2, MMS21 homolog; non-SMC element 2, MMS21 homolog (S. cerevisiae); non-structural maintenance of chromosomes element 2 homolog; NSE2; NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase; NSE2/MMS21 homolog, SMC5-SMC6 complex SUMO ligase; NSMCE2; RGD1305156; Unknown (protein for MGC:133996); zinc finger, MIZ-type containing 7; ZMIZ7
Rabbit
Antigen affinity chromatography
RUO
286053
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96MF7
Nsmce2
Recombinant Human NSMCE2. Recombinant protein control fragment (Product #RP-104928).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.