Learn More
Tocris Bioscience™ NS 19504
Description
NS 19504 is a selective activator of large-conductance Ca2+-activated potassium channels (BKCa, KCa1.1) (EC50 = 11 μM). Activates BK channels in native smooth muscle cells from guinea pig bladder and reduces spontaneous phasic contractions in bladder strips ex vivo. Displays no effect on contractions induced by a high K+ concentration or at 65 other receptors tested, including L- and N- type Ca2+ channels, Nav, KATP and hERG channels.
Specifications
Specifications
| Chemical Name or Material | 5-[(4-Bromophenyl)methyl]-2-thiazolamine |
| CAS | 327062-46-4 |
| Quantity | 10 mg |
| Target | Ca2+-Activated Potassium Channel Activators |
| Molecular Formula | C10H9BrN2S |
| Formula Weight | 269.16 |
| Purity | >99% |