missing translation for 'onlineSavingsMsg'
Learn More

NRF2 NFE2L2 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech

Product Code. 17482633 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17482633 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17482633 Supplier Proteintech Supplier No. CL59416396100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The NRF2, NFE2L2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NRF2, NFE2L2. This antibody recognizes human, mouse, rat antigen. The NRF2, NFE2L2 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.

Specifications

Antigen NRF2 NFE2L2
Applications Immunofluorescence, Flow Cytometry (Intracellular Staining)
Classification Polyclonal
Conjugate CoraLite 594
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 4780
Target Species Human, Rat, Mouse
Isotype IgG
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.