Learn More
Nrf2-IN-1, MedChemExpress
Product Code. 16443238
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16443238
Brand: Medchem Express HY101025
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Nrf2-IN-1 is an inhibitor of nuclear factor-erythroid 2-related factor 2 (Nrf2). Nrf2-IN-1 is developed for the research of acute myeloid leukemia (AML).
Nrf2-IN-1 is an inhibitor of nuclear factor-erythroid 2-related factor 2 (Nrf2). Nrf2-IN-1 is developed for the research of acute myeloid leukemia (AML).
Specifications
Chemical Name or Material Nrf2-IN-1
Physical Form Solid
CAS 1610022-76-8
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C21H22ClN3O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 260 mg/mL (677.31 mM; Need ultrasonic)
SMILES O=C(NO)C1=CC(C2=CC=C(Cl)C=C2)=NN1CC3=CC=C(C(C)(C)C)C=C3
Molecular Weight (g/mol) 383.87
Formula Weight 383.87
Percent Purity 99.89%
Grade Research