Learn More
NRAS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10724-1-AP-20UL
 View more versions of this product
Product Code. 16857293
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
NRAS (GTPase NRas) is a membrane protein that shuttles between the Golgi apparatus and the plasma membrane. This shuttling is regulated through palmitoylation and depalmitoylation by the ZDHHC9-GOLGA7 complex. This protein, which has intrinsic GTPase activity, is activated to a GTP-bound form by a GTPase activating protein and inactivated to a GDP-bound form by a guanine nucleotide-exchange factor. Defects in the gene encoding this protein are a cause of juvenile myelomonocytic leukemia (JMML) and Noo syndrome 6.
Specifications
Specifications
NRAS
Polyclonal
Unconjugated
NRAS
ALPS4, GTPase NRas, HRAS1, N ras, NRAS, NRAS1, Transforming protein N Ras
Rabbit
Antigen Affinity Chromatography
RUO
18176, 24605, 4893
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Flow Cytometry, Immunocytochemistry
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P01111, P08556, Q04970
Nras
NRAS Fusion Protein Ag1081
20 μL
Primary
Human, Rat, Mouse