missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NPY Monoclonal Antibody (8)

Description
ABS 028-08-02 detects Neuropeptide Y in human and rat samples. ABS 028-08-02 has been successfully used in ELISA and immunohistochemistry (paraffin) procedures. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
This gene encodes a neuropeptide that is widely expressed in the central nervous system and influences many physiological processes, including cortical excitability, stress response, food intake, circadian rhythms, and cardiovascular function. The neuropeptide functions through G protein-coupled receptors to inhibit adenylyl cyclase, activate mitogen-activated protein kinase, regulate intracellular calcium levels, and activate potassium channels. A polymorphism in this gene resulting in a change of leucine 7 to proline in the signal peptide is associated with elevated cholesterol levels, higher alcohol consumption, and may be a risk factor for various metabolic and cardiovascular diseases.
Specifications
Specifications
| Antigen | NPY |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 8 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | NPY |
| Gene Accession No. | P01303, P07808, P57774 |
| Gene Alias | 0710005A05Rik; C-flanking peptide of NPY; CPON; involved in appetite regulation; LOC100912228; Neuropeptide tyrosine; Neuropeptide Y; neuropeptide Y precursor; Neuropeptide-Y; Npy; NPY02; OTTHUMP00000201947; prepro-neuropeptide Y; proneuropeptide Y; pro-neuropeptide Y; pro-neuropeptide Y-like; PYY4; RATNPY; RATNPY02 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?